Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: MADKRKLQGE IDRCLKKVSE GVEQFEDIWQ KLHNAANANQ KEKYEADLKK EIKKLQRLRD QIKTWVASNE IKDKRQLIDN RKLIETQMER FKVVERETKT
Component of the CCR4-NOT complex which is one of the major cellular mRNA deadenylases and is linked to various cellular processes including bulk mRNA degradation, miRNA-mediated repression, translational repression during translational initiation and general transcription regulation. Additional complex functions may be a consequence of its influence on mRNA expression. May be involved in metabolic regulation; may be involved in recruitment of the CCR4-NOT complex to deadenylation target mRNAs involved in energy metabolism. Involved in mitotic progression and regulation of the spindle assembly checkpoint by regulating the stability of MAD1L1 mRNA. Can repress transcription and may link the CCR4-NOT complex to transcriptional regulation; the repressive function may involve histone deacetylases. Involved in the maintenance of embryonic stem (ES) cell identity. [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CCR4-associated factor 3; CCR4-NOT transcription complex subunit 3; CCR4-NOT transcription complex, subunit 3; Leukocyte receptor cluster member 2; NOT3 (negative regulator of transcription 3, yeast) homolog
Gene Aliases: CNOT3; KIAA0691; LENG2; NOT3; NOT3H
UniProt ID: (Human) O75175
Entrez Gene ID: (Human) 4849
Molecular Function:
general transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support