Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Immunogen sequence: IDKPSDSLS IGNGDNSQQI SNSDTPSPPP GLSKSNPVIP ISSSNHSARS PFEGAVTESQ SLFSDIFRHP NPIPSGLPPF PSSPQTSSDW PTAPEPQSLF TSETIPVSSS TDWQAAFGFG SSKQPEDDLG FDPFDVTRKA LADLIEKELS VQDQPSLSPT SLQNSSSHTT TAKGPGSGFL HPAAATNANS LNSTFSVLPQ RFPQFQQHRA VYNSFSFPGQ AARYPWMAFP RNSIMHLNHT ANPTSNSNFL DLNLPPQHNT GLGGIPVAGE EEVKVSTMPL STSSHSLQQG QQPTSLHTTV A (273-572 aa encoded by BC035590)
CNOT4 is a 575 amino acid protein with E3 ubiquitin ligase activity, mainly involved in protein modification and protein ubiquitination. It contains a ring type zinc finger domain of the C4C4 type, an RNA recognition motif and a bipartite nuclear localization signal.Studies show that CNOT4 is a component of the CCR4-NOT (CAF1) core complex that functions as general transcription regulation complex and has both a positive as well as a negative effect on the transcription of multiple functionally unrelated genes, thus controls gene expression. CNOT4 interacts with CNOT1 via C-terminus and with E2 ubiquitin ligases via C4C4 RING domain. Reports suggest that CNOT4 gets phosphorylated in response to DNA damage on consensus sites recognized by ATM and ATR.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CCR4-associated factor 4; CCR4-NOT transcription complex subunit 4; CCR4-NOT transcription complex, subunit 4; E3 ubiquitin-protein ligase CNOT4; negative regulator of transcription 4; NOT4 (negative regulator of transcription 4, yeast) homolog; Potential transcriptional repressor NOT4Hp; RING-type E3 ubiquitin transferase CNOT4; transcriptional repressor NOT4Hp
Gene Aliases: CLONE243; CNOT4; NOT4; NOT4H; Not4hp
UniProt ID: (Human) E7ET38, (Mouse) Q8BT14
Entrez Gene ID: (Human) 4850, (Rat) 312227, (Mouse) 53621
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support