Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: MPAATVDHSQ RICEVWACNL DEEMKKIRQV IRKYNYVAMD TEFPGVVARP IGEFRSNADY QYQLLRCNVD LLKIIQLGLT FMNEQGEYPP GTSTWQFNFK FNLTEDMYAQ DSIELLTTSG IQFKKHEEEG IETQYFAELL MTSGVVLCEG VKWLSFHSGY DFGYLIKILT NSNLPEEELD FFEILRLFFP VIYDVKYLMK SCKNLKGGLQ EVAEQLELER IGPQHQAGSD SLLTGMAFFK MREMFFEDHI DDAKYCGHLY GLGSGSSYVQ NGTGNAYEEE ANKQS
CNOT7 (CCR4-NOT transcription complex subunit 7) has 3'-5' poly(A) exoribonuclease activity for synthetic poly(A) RNA substrate. Its function seems to be partially redundant with that of CNOT8. CNOT7 is a catalytic component of the CCR4-NOT complex, which is one of the major cellular mRNA deadenylases, and is linked to various cellular processes including bulk mRNA degradation, miRNA-mediated repression, translational repression during translational initiation, and general transcription regulation. During miRNA-mediated repression, the complex seems to act as a transcriptional repressor during translational initiation, as well. Additional complex functions may be a consequence of its influence on mRNA expression. CNOT7 associates with members of the BTG family such as TOB1 and BTG2 and is required for their anti-proliferative activity.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: BTG1-binding factor 1; CAF-1; Caf1a; carbon catabolite repressor protein (CCR4)-associative factor 1; CCR4-associated factor 1; CCR4-NOT transcription complex subunit 7
Gene Aliases: CAF-1; CAF1; Caf1a; CNOT7; hCAF-1
UniProt ID: (Human) Q9UIV1
Entrez Gene ID: (Human) 29883
Molecular Function:
mRNA polyadenylation factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support