Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 4
Positive Control: Mouse cerebellum tissue lysate, rat brain tissue lysate, NIH/3T3 cell lysate, human fetal skeletal muscle tissue, mouse heart tissue, Hela.
Subcellular Location: Cell membrane.
This gene encodes a hematopoeitin/interferon-class receptor belonging to the cytokine superfamily of receptors. The encoded gene product represents the CNTF-specific alpha subunit of a heterotrimer forming the CNTF receptor complex, which also includes LIFR and gp130. The receptor is attached to the membrane by a glycosyl-phosphatidylinositol linkage and contains an immunoglobulin-like C2-type domain and a fibronectin type-III domain. Signal transduction requires that CNTF bind first to this alpha component, which permits the recruitment of gp130 and LIFR beta to form the tripartite receptor complex. Signal transduction stimulates gene expression, cell survival or differentiation in a variety of neuronal cell types. Alternative splicing has been observed at this locus and two variants, both encoding the same protein, have been identified.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ciliary neurotrophic factor receptor alpha; Ciliary neurotrophic factor receptor subunit alpha; CNTF receptor subunit alpha; CNTFR alpha; CNTFR-alpha; MGC1774
Gene Aliases: CNTFR; Cntfralpha
UniProt ID: (Human) P26992, (Rat) Q08406, (Mouse) O88507
Entrez Gene ID: (Human) 1271, (Rat) 313173, (Mouse) 12804
Molecular Function:
transmembrane signal receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support