Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: LHQVKINREE AVVMVEVNQS AKKQVILSSG TE
Highest antigen sequence identity to the following orthologs: Mouse - 81%, Rat - 81%.
The specific function of this protein remains unknown. The protein encoded by this gene belongs to the NCP family of cell-recognition molecules. This family represents a distinct subgroup of the neurexins. NCP proteins mediate neuron-glial interactions in vertebrates and glial-glial contact in invertebrates. The protein encoded by this gene may play a role in cell recognition within the nervous system. Alternatively spliced transcript variants encoding different isoforms have been described but their biological nature has not been determined.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cell recognition molecule Caspr3; Contactin-associated protein-like 3; RP11-138L21.1; RP11-290L7.1
Gene Aliases: CASPR3; CNTNAP3; CNTNAP3A; KIAA1714
UniProt ID: (Human) Q9BZ76
Entrez Gene ID: (Human) 79937
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support