Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Immunogen sequence: MAINRFRLEN DLEELALYQI QLLKDLRHTE NEEDKVSSSS FRQRMLGNLL RPPYERPELP TCLYVIGLTG ISGSGKSSIA QRLKGLGAFV IDSDHLGHRA YAPGGPAYQP VVEAFGTDIL HKDGIINRKV LGSRVFGNKK QLKILTDIMW PIIAKLAREE MDRAVAEGKR VCVIDAAVLL EAGWQNLVHE VWTAVIPETE AVRRIVERDG LSEAAAQSRL QSQMSGQQLV EQSHVVLSTL WEPHITQRQV EKAWALLQKR IPKTHQALD
Biosynthesis of coenzyme A (CoA) from pantothenic acid (vitamin B5) is an essential universal pathway in prokaryotes and eukaryotes. COASY is a bifunctional enzyme that catalyzes the 2 last steps in CoA synthesis. These activities are performed by 2 separate enzymes, phosphopantetheine adenylyltransferase and dephospho-CoA kinase, in prokaryotes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Bifunctional coenzyme A synthase; bifunctional phosphopantetheine adenylyl transferase/dephospho CoA kinase; CoA synthase; Coenzyme A synthase; NBP; nucleotide binding protein; phosphopantetheine adenylyltransferase / dephosphocoenzyme A kinase; POV-2
Gene Aliases: 1300003G02Rik; COASY; DPCK; NBIA6; NBP; pOV-2; PPAT; PSEC0106; UKR1
UniProt ID: (Human) Q13057, (Mouse) Q9DBL7
Entrez Gene ID: (Human) 80347, (Mouse) 71743, (Rat) 287711
Molecular Function: kinase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support