Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: VGDMLAWLH QATASEKEHL EALLKHVTTQ GVEENIQEVV GHITEGVCRP LKVRIEQVIV AEPGAVLLYK ISNLLKFYHH TISGIVGNSA TALLTTIEEM HLLSKKIFFN SLSLHASKLM DKVELPPPDL GPSSALNQTL MLLREVLASH DSSVVPLDAR QADFVQVLSC VLDPLLQMCT VSASNLGTAD MATFMVNSLY MMKTTLALFE FTDRRLEMLQ FQIEAHLDTL INEQASYVLT RVGLSYIYNT VQQHKPEQGS LANMPNLDSV TLKAAMVQFD RYLSAPDNLL IPQLNFLLSA TVKEQIVKQS TELVCRAYGE VYAAVMNPIN EYKDPENILH RSPQQVQTLL S (308-657 aa encoded by BC051723)
This gene encodes a subunit of the conserved oligomeric Golgi complex that is required for maintaining normal structure and activity of the Golgi apparatus. The encoded protein is organized with conserved oligomeric Golgi complex components 5, 7 and 8 into a sub-complex referred to as lobe B. Alternative splicing results in multiple transcript variants.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: COG complex subunit 6; complexed with Dor1p 2; Component of oligomeric Golgi complex 6; conserved oligomeric Golgi complex protein 6; Conserved oligomeric Golgi complex subunit 6; testicular tissue protein Li 41
Gene Aliases: 4933405E16Rik; AU018618; CDG2L; COD2; COG6; KIAA1134; mKIAA1134; SHNS
UniProt ID: (Human) Q9ULT5, (Mouse) Q8R3I3, (Rat) Q68FP9
Entrez Gene ID: (Human) 57511, (Mouse) 67542, (Rat) 310411
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support