Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
Keep as concentrated solution.
Predicted reactivity: Mouse (89%), Rat (89%), Bovine (93%).
Positive Control: human COLC11A2-transfected 293T cells (partial fragment).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes one of the two alpha chains of type XI collagen, a minor fibrillar collagen. It is located on chromosome 6 very close to but separate from the gene for retinoid X receptor beta. Type XI collagen is a heterotrimer but the third alpha chain is a post-translationally modified alpha 1 type II chain. Proteolytic processing of this type XI chain produces PARP, a proline/arginine-rich protein that is an amino terminal domain. Mutations in this gene are associated with type III Stickler syndrome, otospondylomegaepiphyseal dysplasia (OSMED syndrome), Weissenbacher-Zweymuller syndrome, autosomal dominant non-syndromic sensorineural type 13 deafness (DFNA13), and autosomal recessive non-syndromic sensorineural type 53 deafness (DFNB53). Alternative splicing results in multiple transcript variants. A related pseudogene is located nearby on chromosome 6.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Collagen alpha-2(XI) chain; collagen, type XI, alpha 2; DAQB-79P13.8; HGNC:2187; pro-a2 chain of collagen type XI; procollagen, type XI, alpha 2; type XI collagen alpha2 chain
Gene Aliases: COL11A2; DFNA13; DFNB53; FBCG2; HKE5; PARP; STL3
UniProt ID: (Human) P13942, (Mouse) Q64739
Entrez Gene ID: (Human) 1302, (Mouse) 12815, (Rat) 294279
Molecular Function: extracellular matrix structural protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support