Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Aliquoting is unnecessary for -20°C storage.
COMMD5 is a protein coding gene. May modulate activity of cullin-RING E3 ubiquitin ligase (CRL) complexes. Negatively regulates cell proliferation. Negatively regulates cell cycle G2/M phase transition probably by transactivating p21/CDKN1A through the p53/TP53-independent signaling pathway. Involved in kidney proximal tubule morphogenesis. Down-regulates activation of NF-kappa-B.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: COMM domain-containing protein 5; FLJ13008; HCaRG; hypertension-related calcium regulated; Hypertension-related calcium-regulated gene protein; hypertension-related, calcium regulated
Gene Aliases: COMMD5; HCARG; HT002
UniProt ID: (Human) Q9GZQ3, (Rat) Q9ERR2
Entrez Gene ID: (Human) 28991, (Rat) 245974
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support