Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
COPS8 is a COP9 signalosome (CSN), which is an evolutionarily conserved protein complex of the eight subunits that interacts with deubiquitinating enzymes and protein kinases and is highly homologous to the lid sub-complex of 26S proteasome. The CSN complex is an essential regulator of the ubiquitin conjugation pathway by mediating the deneddylation of the SCF-type E3 ligase complexes, which leads to a decrease in ubiquitin ligase activity of SCF-complexes such as SCF, CSA or DDB2. It is also involved in phosphorylation of p53, c-jun/JUN, ITPK1 and IRF8/ICSBP, possibly via its association with CK2 and PKD kinases. CSN8 encodes the smallest and the least conserved but first identified subunit of CSN. Recent studies show CSN8 is essential for Drosophila development and is essential for peripheral T cell homeostasis and antigen receptor-induced entry into the cell cycle from quiescence.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: COP9 (constitutive photomorphogenic) homolog, subunit 8; COP9 constitutive photomorphogenic homolog subunit 8; COP9 homolog; COP9 signalosome complex subunit 8; COP9 signalosome subunit 8; hCOP9; JAB1-containing signalosome subunit 8; SGN8; signalosome subunit 8
Gene Aliases: 9430009J09Rik; AA408242; COP9; COPS8; CSN8; SGN8
UniProt ID: (Human) Q99627, (Mouse) Q8VBV7, (Rat) Q6P4Z9
Entrez Gene ID: (Human) 10920, (Mouse) 108679, (Rat) 363283
Molecular Function:
scaffold/adaptor protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support