Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MEALILEPSL YTVKAILILD NDGDRLFAKY YDDTYPSVKE QKAFEKNIFN KTHRTDSEIA LLEGLTVVYK SSIDLYFYVI GSSYENELML MAVLNCLFDS LSQMLRKNVE KRALLENMEG LFLAVDEIVD GGVILESDPQ QVVHRVALRG EDVPLTEQTV SQVLQSAKEQ IKWSLLR
This gene encodes a subunit of the cytoplasmic coatamer protein complex, which is involved in autophagy and intracellular protein trafficking. The coatomer protein complex is comprised of seven subunits and functions as the coat protein of coat protein complex (COP)I-vesicles. Alternative splicing results in multiple transcript variants.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: coatomer protein complex, subunit zeta 1; Coatomer subunit zeta-1; nonclathrin coat protein zeta1-COP; Zeta-1 COP; Zeta-1-coat protein
Gene Aliases: 5930435A22Rik; AA407760; CGI-120; COPZ; COPZ1; D4Ertd360e; HSPC181; zeta-COP; zeta1-COP
UniProt ID: (Human) P61923, (Mouse) P61924
Entrez Gene ID: (Human) 22818, (Mouse) 56447
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support