Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Positive test controls include: HeLa, NCI-H460, MCF7, Mouse liver, Mouse brain, Mouse heart, Mouse kidney, Rat liver. The target is usually found in the following locations: Matrix side, Mitochondrion inner membrane, Peripheral membrane protein.
Immunogen sequence: PGVFNEYRTI WFKSYRTIFS CLNRIKSFRY PWARLYSTSQ TTVDSGEVKT FLALAHKWWD EQGVYAPLHS MNDLRVPFIR DNLLKTIPNH QPGKPLLGMK ILDVGCGGGL LTEPLGRLGA SVIGIDPVDE NIKTAQCHKS FDPVLDKRIE YRVCSLEEIV EETAETFDAV VASEVVEHVI DLETFLQCCC QVLKPGGSLF ITTINKTQLS YALGIVFSEQ IASIVPKGTH TWEKFVSPET LESILESNGL SVQTVVGMLY NPFSGYWHWS ENTSLNYAAY AVKSRVQEHP ASAEFVLKGE TEELQANACT NPAVHEKLKK
Ubiquinone, also known as coenzyme Q, or Q, is a critical component of the electron transport pathways of both eukaryotes and prokaryotes. This lipid consists of a hydrophobic isoprenoid tail and a quinone head group. The tail varies in length depending on the organism, but its purpose is to anchor coenzyme Q to the membrane. The quinone head group is responsible for the activity of coenzyme Q in the respiratory chain. The S. cerevisiae COQ3 gene encodes an O-methyltransferase required for 2 steps in the biosynthetic pathway of coenzyme Q. This enzyme methylates an early coenzyme Q intermediate, 3,4-dihydroxy-5-polyprenylbenzoic acid, as well as the final intermediate in the pathway, converting demethyl-ubiquinone to coenzyme Q. The COQ3 gene product is also capable of methylating the distinct prokaryotic early intermediate 2-hydroxy-6-polyprenyl phenol.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 2-polyprenyl-6-hydroxyphenol methylase; 2-polyprenyl-6-hydroxyphenyl methylase; 3,4-dihydroxy-5-hexaprenylbenzoate methyltransferase; 3-demethylubiquinol 3-O-methyltransferase; 3-demethylubiquinone-10 3-methyltransferase; 3-demethylubiquinone-9 3-methyltransferase; Coenzyme Q (ubiquinone); coenzyme Q3 homolog methyltransferase; coenzyme Q3 homolog, methyltransferase; coenzyme Q3 methyltransferase; DHHB methyltransferase; DHHB-MT; DHHB-MTase; dihydroxyhexaprenylbenzoate methyltransferase; hexaprenyldihydroxybenzoate methyltransferase, mitochondrial; methyltransferase COQ3; Polyprenyldihydroxybenzoate methyltransferase; Ubiquinone biosynthesis O-methyltransferase, mitochondrial
Gene Aliases: 4732433J24; bA9819.1; C77934; COQ3; DHHBMT; DHHBMTASE; UG0215E05
UniProt ID: (Human) Q9NZJ6, (Mouse) Q8BMS4, (Rat) Q63159
Entrez Gene ID: (Human) 51805, (Mouse) 230027, (Rat) 29309
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support