Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Promotions
View available promotion(s)
Promo Code: P5877545 Don't miss out: buy 3, only pay for 2 From June 2-13 take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Immunogen sequence: MATLLRPVL RRLCGLPGLQ RPAAEMPLRA RSDGAGPLYS HHLPTSPLQK ALLAAGSAAM ALYNPYRHDM VAVLGETTGH RTLKVLRDQM RRDPEGAQIL QERPRISTST LDLGKLQSLP EGSLGREYLR FLDVNRVSPD TRAPTRFVDD EELAYVIQRY REVHDMLHTL LGMPTNILGE IVVKWFEAVQ TGLPMCILGA FFGPIRLGAQ SLQVLVSELI PWAVQNGRRA PCVLNLYYER RWEQSLRALR EELGITAPPM HVQGLA (1-265 aa encoded by B C011895)
Component of the coenzyme Q biosynthetic pathway. May play a role in organizing a multi-subunit COQ enzyme complex required for coenzyme Q biosynthesis. Required for steady-state levels of other COQ polypeptides.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Coenzyme Q biosynthesis protein 4 homolog; coenzyme Q biosynthesis protein 4 homolog {ECO:0000255|HAMAP-Rule:MF_03111}; coenzyme Q4 homolog; EST-MNCb4625; Ubiquinone biosynthesis protein COQ4 homolog, mitochondrial; ubiquinone biosynthesis protein COQ4 homolog, mitochondrial {ECO:0000255|HAMAP-Rule:MF_03111}
Gene Aliases: A330041N06; CGI-92; COQ10D7; COQ4; D2Ertd97e
UniProt ID: (Human) Q9Y3A0, (Mouse) Q8BGB8, (Rat) Q4FZU1
Entrez Gene ID: (Human) 51117, (Mouse) 227683, (Rat) 366013
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support