Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 4
This antibody is predicted to react with bovine and rat based on sequence homology.
Methyltransferase required for the conversion of 2-polyprenyl-6-methoxy-1,4-benzoquinol (DDMQH2) to 2-polyprenyl-3-methyl-6-methoxy-1,4-benzoquinol (DMQH2).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 2-methoxy-6-polyprenyl-1,4-benzoquinol methylase, mitochondrial; coenzyme Q5 homolog, methyltransferase; Ubiquinone biosynthesis methyltransferase COQ5; ubiquinone biosynthesis methyltransferase COQ5, mitochondrial
Gene Aliases: 1810014G04Rik; COQ5; D5Ertd33e
UniProt ID: (Human) Q5HYK3, (Mouse) Q9CXI0
Entrez Gene ID: (Human) 84274, (Mouse) 52064
Molecular Function: methyltransferase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support