Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Promotions
View available promotion(s)
Promo Code: P5906365 Don't miss out: buy 3, only pay for 2 Extended to June 27: take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Immunogen sequence: MGGLWRPGW RCVPFCGWRW IHPGSPTRAA ERVEPFLRPE WSGTGGAERG LRWLGTWKRC SLRARHPALQ PPRRPKSSNP FTRAQEEERR RQNKTTLTYV AAVAVGMLGA SYAAVPLYRL YCQTTGLGGS AVAGHASDKI ENMVPVKDRI IKISFNADVH ASLQWNFRPQ QTEIYVVPGE TALAFYRVKN PTDKPVIGIS TYNIVPFEAG QYFNKIQCFC FEEQRLNPQE EVDMPVFFYI DPEFAEDPRM IKVDLITLSY TFFEAKEGHK LPVPGYN (1-276 aa encoded by BC005895)
Cytochrome c oxidase (COX) is the terminal enzyme in the electron transfer chain, functioning as a transmembrane proton pump that builds an electrochemical gradient with chemical energy from the reduction of O2. Cytochrome c oxidase assembly protein COX11 is an intracellular mitochondrial membrane protein necessary for the construction of an active COX complex. COX11 contains a single transmembrane helix downstream of the N-terminal, mitochondrial targeting sequence and a C-terminal Cu(I)-binding domain. The assembly of COX requires the delivery of metal cofactors. Along with COX12 and SCO1/2, COX11 acts as a metal ion chaperone necessary for copper insertion into CuA and CuB redox-active copper centers of COX in eukaryotes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: COX11 cytochrome c oxidase copper chaperone; COX11 homolog, cytochrome c oxidase assembly protein; cytochrome c oxidase assembly homolog 11; cytochrome c oxidase assembly protein 11; Cytochrome c oxidase assembly protein COX11, mitochondrial; cytochrome c oxidase subunit 11
Gene Aliases: 2010004I09Rik; COX11; COX11P
UniProt ID: (Human) Q9Y6N1, (Mouse) Q6P8I6
Entrez Gene ID: (Human) 1353, (Mouse) 69802, (Rat) 690300
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support