Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: SSGAHGEEGS ARMWKTLTFF VALPGVAVSM LNVYLKSHHG EHERPEFIAY PHLRIRTKPF PWGDGNHTLF HNPHVNPLPT GYEDE; Positive Samples: A-549, MCF7, Mouse kidney, Mouse heart, Mouse liver, Rat liver, Rat brain; Cellular Location: Mitochondrion inner membrane
This protein is one of the nuclear-coded polypeptide chains of cytochrome c oxidase, the terminal oxidase in mitochondrial electron transport.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: COX VIa-L; Cytochrome c oxidase polypeptide VIa-liver; Cytochrome c oxidase subunit 6A1, mitochondrial; Cytochrome c oxidase subunit VIa (liver); cytochrome C oxidase subunit VIa homolog; cytochrome c oxidase subunit VIa polypeptide 1; Cytochrome c oxidase subunit VIA-liver; subunit VIaL (liver-type)
Gene Aliases: CMTRID; COX6A; COX6A1; COX6AL; VIaL
UniProt ID: (Human) P12074, (Mouse) P43024, (Rat) P10818
Entrez Gene ID: (Human) 1337, (Mouse) 12861, (Rat) 25282
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support