Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This target displays homology in the following species: Cow: 92%; Guinea Pig: 86%; Horse: 86%; Human: 100%; Mouse: 77%; Rabbit: 79%; Rat: 83%
This protein is one of the nuclear-coded polypeptide chains of cytochrome c oxidase, the terminal oxidase in mitochondrial electron transport.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: COX7AL2; cytochrome c oxidase polypeptide 7A2, mitochondrial; cytochrome c oxidase polypeptide VIIa-liver/heart; Cytochrome c oxidase subunit 7A2, mitochondrial; cytochrome c oxidase subunit VIIa polypeptide 2 (liver); Cytochrome c oxidase subunit VIIa-L; Cytochrome c oxidase subunit VIIa-liver/heart; cytochrome c oxidase subunit VIIaL; hepatic cytochrome-c oxidase chain VIIa
Gene Aliases: COX7A2; COX7AL; COX7AL1; COXVIIa-L; COXVIIAL; VIIAL
UniProt ID: (Human) P14406
Entrez Gene ID: (Human) 1347
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support