Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Sequence of this protein is as follows: SKLPPNHEDL AKVAKIGTDV LSTRYETRYI YGPIESTIYP ISGSSLDWAY DLGIKHTFAF ELRDKGKFGF LLPESRIKPT CRETMLAVKF IAKYILKHTS
Three different forms of human pancreatic procarboxypeptidase A have been isolated. This gene encodes a form which is obtained as a binary complex of a procarboxypeptidase A with proproteinase E and functions as a secretory granule metalloexopeptidase.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Carboxypeptidase A3; carboxypeptidase A3 (mast cell); Mast cell carboxypeptidase A; MC-CPA; tissue carboxypeptidase A
Gene Aliases: CPA3; MC-CPA
UniProt ID: (Human) P15088
Entrez Gene ID: (Human) 1359
Molecular Function: metalloprotease
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support