Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MLFPTSAQES SRGLPDANDL CLGLQSLSLT GWDRPWSTQD SDSSAQSSTH SVLSMLHNPL GNVLGKPPLS FLPLDPLGSD LVDKFPAPSV RGSRLDTRPI LDSRSSSPSD SDTSGFSSGS DHLSDLISSL RISPPLPFLS LSGGGPRDPL KMGVGSRMDQ EQAALAAVTP SPTSASKRWP GASVWPSWDL LEAPKDPFSI EREARLHRQA AAVNEATCTW SGQLPPRNYK NPIYSCKVFL GGVPWDITEA GLVNTFRVFG SLSVEWPGKD GKHPRCPPKG YVYLVFELEK SVRSLLQACS HDPLSPDGLS EYYFKMSSRR MRCKEVQVIP WVLADSNFVR SPSQRLDPSR TVFVGALHGM LNAEALAAIL NDLFGGVVYA GIDTDKHKYP IGSGRVTFNN QRSYLKAVSA AFVEIKTTKF TKKVQIDPYL EDSLCHICSS QPGPFFCRDQ VCFKYFCRSC WHWRHSMEGL RHHSPLMRNQ KNRDSS
This gene encodes a protein which, with 5-lipoxygenase, is required for leukotriene synthesis. Leukotrienes are arachidonic acid metabolites which have been implicated in various types of inflammatory responses, including asthma, arthritis and psoriasis. This protein localizes to the nuclear membrane. Inhibitors of its function impede translocation of 5-lipoxygenase from the cytoplasm to the nuclear membrane and inhibit 5-lipoxygenase activation. Alternatively spliced transcript variants encoding different isoforms have been identified for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CEBP; CPE-binding protein 1; CPE-BP1; Cytoplasmic polyadenylation element-binding protein 1; FLJ13203; h-CEBP; mCPEB-1; MGC34136; MGC60106
Gene Aliases: AU024112; CPE-BP1; CPEB; CPEB-1; CPEB1; h-CPEB; hCPEB-1; mCPEB
UniProt ID: (Human) Q9BZB8, (Mouse) P70166
Entrez Gene ID: (Human) 64506, (Mouse) 12877
Molecular Function: mRNA polyadenylation factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support