Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Aliquoting is unnecessary for -20°C storage.
Proteins encoded by the complexin/synaphin gene family are cytosolic proteins that function in synaptic vesicle exocytosis. These proteins bind syntaxin, part of the SNAP receptor. The protein product of this gene binds to the SNAP receptor complex and disrupts it, allowing transmitter release. Two transcript variants encoding the same protein have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 921-L; Complexin II; Complexin-2; CPX II; CPX-II; DKFZp547D155; MGC138492; synaphin 1; Synaphin-1
Gene Aliases: 921-L; AI413745; AW492120; CPLX2; CPX-2; CPX2; Hfb1
UniProt ID: (Human) Q6PUV4, (Rat) P84087, (Mouse) P84086
Entrez Gene ID: (Pig) 110259396, (Human) 10814, (Rat) 116657, (Mouse) 12890
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support