Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MSDAEAGGVF HRARGRTLAA FPAEKESEWK GPFYFILGAD PQFGLIKAWS TGDCDNGGDE WEQEIRLTEQ AVQAINKLNP KPKFFVLCGD LIHAMP
Highest antigen sequence identity to the following orthologs: Mouse - 82%, Rat - 83%.
Protein phosphatase that dephosphorylates AKT family kinase specifically at 'Ser-473', blocking cell cycle progression and promoting cell apoptosis. May play an inhibitory role in glucose uptake by adipocytes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: calcineurin-like phosphoesterase domain containing 1; Calcineurin-like phosphoesterase domain-containing protein 1; complete S transactivated protein 1; Complete S-transactivated protein 1; Serine/threonine-protein phosphatase CPPED1
Gene Aliases: CPPED1; CSTP1
UniProt ID: (Human) Q9BRF8
Entrez Gene ID: (Human) 55313
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support