Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This antibody can bind the close sequences genes.
Immunogen sequence: SFTLIVFSN GKLGLSVEHS WADCPISGHM WEFTLATECF QLGYSTDGHC KGHPDPTLPQ PQRLQWDLPD QIHSSISLAL RGAKILSENV DCHVVPFSLF GKSFIRRCHL SSDSFIQIAL QLAHFRDRGQ FCLTYESAMT RLFLEGRTET VRSCTREACN FVRAMEDKEK TDPQCLALFR VAVDKHQALL KAAMSGQGVD RHLFALYIVS RFLHLQSPFL TQVHSEQWQL STSQIPVQQM HLFDVHNYPD YVSSGGGFGP ADDHGYGVSY IFMGDGMITF HISSKKSSTK TDSHRLGQHI EDALLDVASL FQAGQHFKRR FRGSGKENSR HRCGFLSRQT GASKASMTST DF (442-793 aa encoded by BC029104)
This gene encodes a member of the carnitine/choline acetyltransferase family. The encoded protein regulates the beta-oxidation and transport of long-chain fatty acids into mitochondria, and may play a role in the regulation of feeding behavior and whole-body energy homeostasis. Alternatively spliced transcript variants encoding multiple protein isoforms have been observed for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Carnitine O-palmitoyltransferase 1, brain isoform; carnitine O-palmitoyltransferase I, brain isoform; Carnitine palmitoyltransferase 1C; Carnitine palmitoyltransferase I; carnitine palmitoyltransferase I related C; CPT I-C; CPTI-B; FLJ23809; Palmitoyl thioesterase CPT1C
Gene Aliases: CATL1; CPT1-B; CPT1C; CPT1P; CPTI-B; CPTIC; SPG73
UniProt ID: (Human) Q8TCG5
Entrez Gene ID: (Pig) 100519258, (Human) 126129
Molecular Function:
acyltransferase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support