Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: WFDKSFNLII AKDGSTAVHF EHSWGDGVAV LRFFNEVFKD STQTPAVTPQ SQPATTDSTV TVQKLNFELT DALKTGITAA KEKFDATMKT LTIDCVQFQR
Carnitine palmitoyltransferase II precursor (CPT2) is a nuclear protein which is transported to the mitochondrial inner membrane. CPT2 together with carnitine palmitoyltransferase I oxidizes long-chain fatty acids in the mitochondria. Defects in this gene are associated with mitochondrial long-chain fatty-acid (LCFA) oxidation disorders.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Carnitine O-palmitoyltransferase 2, mitochondrial; Carnitine palmitoyltransferase II; CPT II; OTTHUMP00000010493; testicular secretory protein Li 13
Gene Aliases: CPT1; CPT2; CPTASE; IIAE4
UniProt ID: (Human) P23786
Entrez Gene ID: (Human) 1376
Molecular Function: acyltransferase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support