Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Immunogen sequence: LRHLAAAKGI ILPELYLDPA YGQINHNVLS TSTLSSPAVN LGGFAPVVSD GFGVGYAVHD NWIGCNVSSY PGRNAREFLQ CVEKALEDMF DALEGKSIKS
Carnitine palmitoyltransferase II precursor (CPT2) is a nuclear protein which is transported to the mitochondrial inner membrane. CPT2 together with carnitine palmitoyltransferase I oxidizes long-chain fatty acids in the mitochondria. Defects in this gene are associated with mitochondrial long-chain fatty-acid (LCFA) oxidation disorders.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Carnitine O-palmitoyltransferase 2, mitochondrial; Carnitine palmitoyltransferase II; CPT II; OTTHUMP00000010493; testicular secretory protein Li 13
Gene Aliases: AI323697; Cpt-2; CPT1; CPT2; CPTASE; CPTII; IIAE4
UniProt ID: (Human) P23786, (Mouse) P52825, (Rat) P18886
Entrez Gene ID: (Human) 1376, (Mouse) 12896, (Rat) 25413
Molecular Function: acyltransferase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support