Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Antibody detects endogenous levels of total CPXCR1.
The CPXCR1 gene is not extensively covered in the scientific literature. CXCL1, CXCR1, and CXCR2 are involved in chemotactic cytokine activities, which are crucial in inflammatory responses and conditions. CXCL1, for example, plays significant roles in angiogenesis and the recruitment of neutrophils under inflammatory conditions. Although CPXCR1 might be related due to its nomenclature and potential function in chemokine activity.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cancer/testis antigen 77; CPX chromosomal region candidate gene 1 protein; CT77; hypothetical LOC100360571
Gene Aliases: CPXCR1; CT77
UniProt ID: (Human) Q8N123
Entrez Gene ID: (Human) 53336, (Rat) 100360571
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support