Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
Immunogen sequence: MPNFAGTWK MRSSENFDEL LKALGVNAML RKVAVAAASK PHVEIRQDGD QFYIKTSTTV RTTEINFKVG EGFEEETVDG RKCRSLATWE NENKIHCTQT LLEGDGPKTY WTSELANDEL ILTFGADDVV CTRIYVRE (1-137 aa encoded by BC022069)
This gene encodes a specific binding protein for a vitamin A family member and is thought to play an important role in retinoic acid-mediated differentiation and proliferation processes. It is structurally similar to the cellular retinol-binding proteins, but binds only retinoic acid at specific sites within the nucleus, which may contribute to vitamin A-directed differentiation in epithelial tissue.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: cellular retinoic acid binding protein I; Cellular retinoic acid-binding protein 1; Cellular retinoic acid-binding protein I; CRABP-I
Gene Aliases: AI326249; CRABP; Crabp-1; CRABP-I; CRABP1; CRABPI; Rbp-5; RBP5
UniProt ID: (Human) P29762, (Rat) P62966, (Mouse) P62965
Entrez Gene ID: (Human) 1381, (Rat) 25061, (Mouse) 12903
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support