Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 7
Recommended positive controls: A549, HepG2, mouse brain, mouse fetal brain, rat brain.
Predicted reactivity: Mouse (94%), Rat (94%), Xenopus laevis (89%), Pig (95%), Rhesus Monkey (100%), Bovine (97%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Cellular retinoic acid binding protein (CRABP) is a low molecular weight protein with unknown function and existing in two isoforms, CRABP1 and CRABP2. The predicted amino acid sequences of human CRABP1 and CRABP2 exhibits 99.3 and 93.5% identity to mouse CRABP1 and CRABP2, respectively. The CRABP2 isoform is important in retinoic acid-mediated regulation of human skin growth and differentiation. The CRABP2 gene localized on human chromosome 1q21.3 is transcriptionally regulated by a newly synthesized regulatory protein. The protein is expressed in human adult liver.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: cellular lipid-binding protein; cellular retinoic acid binding protein II; Cellular retinoic acid-binding protein 2; Cellular retinoic acid-binding protein II; CRABP-II; RP11-66D17.4
Gene Aliases: AI893628; Crabp-2; CRABP-II; CRABP2; CrabpII; RBP6
UniProt ID: (Human) P29373, (Mouse) P22935, (Rat) P51673
Entrez Gene ID: (Human) 1382, (Mouse) 12904, (Rat) 29563
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support