Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
This antibody recognizes both of CRACR2A-c and CRACR2A-a.
Immunogen sequence: MAAPDGRVV SRPQRLGQGS GQGPKGSGAC LHPLDSLEQK ETQEQTSGQL VMLRKAQEFF QTCDAEGKGF IARKDMQRLH KELPLSLEEL EDVFDALDAD GNGYLTPQEF TTGFSHFFFS QNNPSQEDAG EQVAQRHEEK VYLSRGDEDL GDMGEDEEAQ FRMLMDRLGA QKVLEDESDV KQLWLQLKKE EPHLLSNFED FLTRIISQLQ EAHEEKNELE CALKRKIAAY DEEIQHLYEE MEQQIKSEKE QFLLKDTERF QARSQELEQK LLCKEQELEQ LTQKQKRLEG QCTALHHDKH ETKAENTKLK LTNQELAREL ERTSWELQDA QQQLESLQQE ACKLHQEKEM EVYRVTESLQ REKAGLLKQL DFLRCVGGHW PVLRAPPRSL GSEGPV (1-395 aa encoded by B C004524)
CRACR2A is a Ca(2+)-binding protein that plays a key role in store-operated Ca(2+) entry (SOCE) in T-cells by regulating CRAC channel activation. CRACR2A acts as a cytoplasmic calcium-sensor that facilitates the clustering of ORAI1 and STIM1 at the junctional regions between the plasma membrane and the endoplasmic reticulum upon low Ca(2+) concentration. Further, CRACR2A regulates CRAC channel activation, including translocation and clustering of ORAI1 and STIM1. Upon increase of cytoplasmic Ca(2+) resulting from opening of CRAC channels, CRACR2A dissociates from ORAI1 and STIM1, thereby destabilizing the ORAI1-STIM1 complex.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Ca2+ release-activated Ca2+ (CRAC) channel regulator 2A; Calcium release-activated calcium channel regulator 2A; Calcium release-activated channel regulator 2A; CRAC channel regulator 2A; CRAC regulator 2A; EF-hand calcium binding domain 4B; EF-hand calcium-binding domain-containing protein 4B; Ras-related protein Rab-46
Gene Aliases: CRACR2A; EFCAB4B; RAB46
UniProt ID: (Human) Q9BSW2
Entrez Gene ID: (Human) 84766
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support