Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
FIGURE: 1 / 2
Antibody detects endogenous levels of total CREB3L4.
cAMP response element-binding (CREB) proteins are a familyof mammalian transcription activators. For further backgroundinformation on CREB proteins, see CREB1 (MIM 123810).[supplied byOMIM].
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: AIbZIP; Androgen-induced basic leucine zipper protein; androgen-induced bZIP protein; ATCE1; Attaching to CRE-like 1; cAMP responsive element binding protein 1; cAMP responsive element binding protein 3-like 4; cAMP-responsive element-binding protein 3-like protein 4; cAMP-responsive element-binding protein 4; CREB-4; Cyclic AMP-responsive element-binding protein 3-like protein 4; Cyclic AMP-responsive element-binding protein 4; hJAL; mJAL; OTTHUMP00000035266; RP11-422P24.8; Tisp40; Transcript induced in spermiogenesis protein 40
Gene Aliases: 1700012K17Rik; 5330432F22Rik; acre1; AIBZIP; ATCE1; AV040530; AV258827; CREB3; CREB3L4; CREB4; hJAL; JAL; TISP40
UniProt ID: (Human) Q8TEY5, (Mouse) Q9D2A5, (Rat) Q5UEM7
Entrez Gene ID: (Human) 148327, (Mouse) 78284, (Rat) 310616
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support