Search Thermo Fisher Scientific
- ์ฃผ๋ฌธ ํํฉ
- ๋น ๋ฅธ์ฃผ๋ฌธ
-
ํ์์ด ์๋์ ๊ฐ์? ๊ณ์ ์์ฑํ๊ธฐ
Search Thermo Fisher Scientific
FIGURE: 1 / 4
Reconstitution: 1 X 25 ยตL double distilled water (DDW), depending on the sample size. The antibody ships as a lyophilized powder at room temperature. Upon arrival, it should be stored at -20C. The reconstituted solution can be stored at 4C for up to 1 week. For longer periods, small aliquots should be stored at -20C. Avoid multiple freezing and thawing. Centrifuge all antibody preparations before use (10000 x g 5 min).
Corticotropin-releasing factor receptor 1 (CRFR1/CRHR1), a Corticotropin-Releasing Factor Receptor, is involved in mediating the stress response and anxiety-related behaviors. Mutations of the receptor reduce anxiety-related behaviors and may be involved in the pathogenesis of Cushing disease. Multiple isoforms of CRFR1 are produced by alternative splicing. CRFR1 has been reported to be expressed in the pituitary, adrenal gland, brain, colon, ovary, placenta, testis, white blood cells, and skin. ESTs have been isolated from human brain, eye, colon, and embryo libraries.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: corticotrophin releasing hormone receptor 1; corticotropin releasing hormone 1; Corticotropin-releasing factor receptor 1; corticotropin-releasing factor type 1 receptor; Corticotropin-releasing hormone receptor 1; CRF 1 receptor; Crf receptor 1; Crf type 1; CRF-R; CRF-R-1; CRF-R1; CRF-R1alpha; CRF1; Crf1 receptor; CRFR-1; CRH receptor 1; CRH receptor subtype-1 isoform-3; CRH-R 1; CRH-R-1; CRH-R1; CRH-R1h; CRHR1f; seven transmembrane helix receptor
Gene Aliases: CRF-R; CRF-R-1; CRF-R1; CRF1; CRFR; CRFR-1; CRFR1; CRH-R-1; CRH-R1; CRHR; CRHR1; CRHR1L
UniProt ID: (Human) P34998, (Rat) P35353, (Mouse) P35347
Entrez Gene ID: (Human) 1394, (Rat) 58959, (Mouse) 12921
Molecular Function:
transmembrane signal receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support