Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
FIGURE: 1 / 5
Antibody detects endogenous levels of total Gliomedin.
CRG-L2 is a protein that contains olfactomedin-like and collagen-like domains. The encoded protein, which exists in both transmembrane and secreted forms, promotes formation of the nodes of Ranvier in the peripheral nervous system. Mutations in this gene cause a form of lethal congenital contracture syndrome in human patients. Autoantibodies to the encoded protein have been identified in sera form patients with multifocal motor neuropathy.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: cancer related gene - liver 2; Cancer-related gene liver 2 protein; collomin; colmedin; CRG-L2; FLJ23917; Gliomedin; olfactomedin-related protein
Gene Aliases: CLOM; COLM; CRG-L2; CRGL2; Crlg2; GLDN; UNC-112; UNQ9339/PRO34011
UniProt ID: (Human) Q6ZMI3, (Rat) Q80WL1, (Mouse) Q8BMF8
Entrez Gene ID: (Human) 342035, (Rat) 315675, (Mouse) 235379
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support