Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Corticotropin-releasing hormone (CRH; 122560) is a 41-amino acid peptide synthesized in the hypothalamus, and play a role in the neuroregulation, responses to stress and immune challenge. Corticotropin Releasing Factor Receptor 2 (CRHR2) was the receptor of CRH. The sepression of CRHR2 has been reported in various regions of the brain as well as in placenta, umbilical vein, heart, epididymis, gastrointestinal tract, adrenal and skeletal muscle.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: corticotrophin releasing hormone receptor 2; corticotropin releasing factor receptor type 2 beta; corticotropin-releasing factor 2; Corticotropin-releasing factor receptor 2; corticotropin-releasing factor receptor subtype 2; Corticotropin-releasing hormone receptor 2; CRF 2 receptor; CRF-R 2; CRF-R-2; CRF-R2; CRF-RB; CRF2; CRF2 receptor, beta isoform; CRFR-2; CRH receptor 2 variant B; CRH-R 2; CRH-R-2; CRH-R2; Crh2 receptor
Gene Aliases: CRF-R2; CRF-RB; CRF2; CRF2R; CRFR2; CRFR2alpha; CRFR2beta; CRH2R; CRHR2; HM-CRF
UniProt ID: (Human) Q13324, (Rat) P47866, (Mouse) Q60748
Entrez Gene ID: (Human) 1395, (Rat) 64680, (Mouse) 12922
Molecular Function: transmembrane signal receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support