Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotions
View available promotion(s)
Promo Code: CI2Z9E .
A suggested positive control is Jurkat cell lysate.
PA5-34411 can be used with blocking peptide PEP-1453.
CRIM2, also known as Kielin/chordin-like protein (KCP), is a novel enhancer of BMP signaling. It binds to BMP7, a morphogen that is important for kidney development and part of the kidney's physiological response to repair of acute kidney injury, enhancing its binding to its receptor. Because of this, CRIM2 has been suggested as a potential antifibrotic therapy in acute and chronic kidney injury. CRIM2 can also interact directly with either activin or TGF-Beta1, suppressing activin- and TGF-Beta1-dependent transcription.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CRIM-2; cysteine rich BMP regulator 2 (chordin like); cysteine rich BMP regulator 2 (chordin-like); Cysteine-rich BMP regulator 2; Cysteine-rich motor neuron 2 protein; KCP-1; kielin; Kielin/chordin-like protein; Kielin/chordin-like protein 1
Gene Aliases: AW060220; CRIM-2; CRIM2; Gm793; KCP; KCP-1; KCP1; NET67; RGD1561119
UniProt ID: (Human) Q6ZWJ8, (Mouse) Q3U492
Entrez Gene ID: (Human) 375616, (Rat) 296952, (Mouse) 333088
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support