Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Promotions
View available promotion(s)
Promo Code: P5877545 Don't miss out: buy 3, only pay for 2 Extended to June 20: take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Immunogen sequence: MVCEKCEKK LGTVITPDTW KDGARNTTES GGRKLNENKA LTSKKARFDP YGKNKFSTCR ICKSSVHQPG SHYCQGCAYK KGICAMCGKK VLDTKNYKQT SV (1-101 aa encoded by BC018653)
Involved in the cytoskeletal anchoring of DLG4 in excitatory synapses.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cysteine-rich interactor of PDZ three; Cysteine-rich interactor of PDZ3; Cysteine-rich PDZ-binding protein; postsynaptic protein CRIPT
Gene Aliases: 1200020A08Rik; AA589469; CRIPT; HSPC139; SSMDF
UniProt ID: (Human) Q9P021, (Mouse) O70333, (Rat) Q792Q4
Entrez Gene ID: (Human) 9419, (Mouse) 56724, (Rat) 56725
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support