Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MPAGRRGPAA QSARRPPPLL PLLLLLCVLG APRAGSGAHT AVISPQDPTL LIGSSLLATC SVHGDPPGAT AEGLYWTLNG RRLPPELSRV LNASTLALAL ANLNGSRQRS GDNLVCHARD GSILAGSCLY VGLPPEKPVN ISCWSKNMKD LTCRWTPGAH GETFLHTNYS LKYKLRWYGQ DNTCEEYHTV GPHSCHIPKD LALFTPYEIW VEATNRLGSA RSDVLTLDIL DVVTTDPPPD VHVSRVGGLE DQLSVRWVSP PALKDFLFQA KYQIRYRVED SVDWKVVDDV SNQTSCRLAG LKPGTVYFVQ VRCNPFGIYG SKKAGIWSEW SHPTAASTPR SERPGPGGGA CEPRGGEPSS GPVRRELKQF LGWLKKHAYC SNLSFRLYDQ WRAWMQKSHK TRNQDEGILP SGRRGTARGP AR
CRLF1 encodes a member of the cytokine type I receptor family. The protein forms a secreted complex with cardiotrophin-like cytokine factor 1 and acts on cells expressing ciliary neurotrophic factor receptors. The complex can promote survival of neuronal cells. Mutations in this gene result in Crisponi syndrome and cold-induced sweating syndrome.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: class I cytokine receptor; CLF-1; Cytokine receptor-like factor 1; cytokine type 1 receptor CRLP-1; Cytokine-like factor 1; ZcytoR5
Gene Aliases: CISS; CISS1; CLF; CLF-1; CRLF1; NR6; UNQ288/PRO327; zcytor5
UniProt ID: (Human) O75462
Entrez Gene ID: (Human) 9244
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support