Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
May play a role in the negative regulation of cell cycle progression.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CREME-9; Cytokine receptor-like factor 3; Cytokine receptor-like molecule 9; Cytokine receptor-related factor 4; Cytokine receptor-related protein 4; Type I cytokine receptor-like factor p48
Gene Aliases: BB164954; CREME-9; CREME9; CRLF3; CRLM9; CYTOR4; FRWS; P48; p48.2
UniProt ID: (Human) Q8IUI8, (Mouse) Q9Z2L7
Entrez Gene ID: (Human) 51379, (Rat) 54395, (Mouse) 54394
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support