Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 6
Reconstitution: 25 µL, 50 µL or 0.2 mL double distilled water (DDW), depending on the sample size. The antibody ships as a lyophilized powder at room temperature. Upon arrival, it should be stored at -20C. The reconstituted solution can be stored at 4C, protected from the light, for up to 1 week. For longer periods, small aliquots should be stored at -20C. Avoid multiple freezing and thawing. Centrifuge all antibody preparations before use (10000 X g 5 min).
CRLR is a receptor for calcitonin gene-related peptide type 1. The activity of CRLR is mediated by G proteins which activate adenylyl cyclase. CRLR expression has been reported in brain, lung, blood vessel, liver, and intestinal tract. ESTs have been isolated from B-cell/lung/testis, bome marrow, embryo, lung and synovium libraries.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Calcitonin gene-related peptide type 1 receptor; Calcitonin receptor-like receptor; CGRP receptor; CGRP type 1 receptor; Cgrp1 receptor; CRLR
Gene Aliases: AV071593; CALCRL; CGRPR; CLR; CRLR; RATCRLR; RNCLR
UniProt ID: (Human) Q16602, (Rat) Q63118, (Mouse) Q9R1W5
Entrez Gene ID: (Human) 10203, (Rat) 25029, (Mouse) 54598
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support