Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Antibody detects endogenous levels of total CRLS1.
The function of this protein is still unknown. The gene of human hypothetical protein (LOC54675) is up-regulated in colon cancer metastasis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: cardiolipin synthase; Cardiolipin synthase (CMP-forming); cardiolipin synthetase; CLS; Protein GCD10 homolog
Gene Aliases: 0610009I22Rik; 4930557M15Rik; 5730490M08Rik; C20orf155; CLS; CLS1; CRLS1; dJ967N21.6; GCD10; RGD1311037
UniProt ID: (Human) Q9UJA2, (Mouse) Q80ZM8, (Rat) Q5U2V5
Entrez Gene ID: (Human) 54675, (Mouse) 66586, (Rat) 366196
Molecular Function:
transferase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support