Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Positive test controls include: C6.
Immunogen sequence: MSFQGKKSIP RITSDRLLIR GGRIVNDDQS FYADVHVEDG LIKQIGENLI VPGGIKTIDA HGLMVLPGGV DVHTRLQMPV LGMTPADDFC QGTKAALAGG
Collapsin response mediator proteins (CRMPs), including CRMP-1 (DRP-1), CRMP-2 (DRP-2 or TOAD64), CRMP-3 (DRP-4), CRMP-4 (DRP-3) and CRMP-5 (DRP-5), mediate signal transduction after exposure of neural cells to the axon guidance molecule Semaphorin 3A (SEMA3A)/collapsin. CRMPs are present in the developing cerebral cortex and neocortical neurons and are responsive to SEMA3A. In the adult brain, the expression of CRMPs is dramatically downregulated. However, they remain expressed in structures that retain their capacity for differentiation and plasticity. CRMP-3 is expressed transiently in developing spinal cord and selectively in the postnatal cerebellum.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Collapsin response mediator protein 3; CRMP-3; Dihydropyrimidinase-related protein 4; DRP-4; ULIP-4; ULIP4 protein; UNC33-like phosphoprotein 4
Gene Aliases: Crmp3; Dpys4; Dpysl4; Ulip4
UniProt ID: (Rat) Q62951
Entrez Gene ID: (Rat) 25417
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support