Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This antibody is predicted to react with rat and dog based on sequence homology.
This antibody is tested in Peptide ELISA: antibody detection limit dilution 32,000.
CROT encodes a member of the carnitine/choline acetyltransferase family. The encoded protein converts 4, 8-dimethylnonanoyl-CoA to its corresponding carnitine ester. This transesterification occurs in the peroxisome and is necessary for transport of medium- and long- chain acyl-CoA molecules out of the peroxisome to the cytosol and mitochondria. The protein thus plays a role in lipid metabolism and fatty acid beta-oxidation. Alternatively spliced transcript variants have been described.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: COT; peroxisomal carnitine acyltransferase; Peroxisomal carnitine O-octanoyltransferase
Gene Aliases: 1200003H03Rik; COT; CROT
UniProt ID: (Human) Q9UKG9, (Mouse) Q9DC50
Entrez Gene ID: (Human) 54677, (Mouse) 74114
Molecular Function:
acyltransferase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support