Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
The CRTAM gene is upregulated in CD4-positive and CD8-positive T cells and encodes a type I transmembrane protein with V and C1-like Ig domains.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CD355; CD355 antigen; class I MHC restricted T cell associated molecule; class-I MHC-restricted T cell associated molecule; Class-I MHC-restricted T-cell-associated molecule; cytotoxic and regulatory T cell molecule; Cytotoxic and regulatory T-cell molecule
Gene Aliases: CD355; CRTAM
UniProt ID: (Human) O95727
Entrez Gene ID: (Human) 56253
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support