Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: QDPYGGGGQS AWPAPYMGFC DGENNGHGEV ASFPGPLKEE NLLNVPKPLP KQLWETKEIQ SLSGRPRSCD VGGGNAFPHN GQNLGLSPFL GTLNTGGSL
The protein encoded by this gene has similarity to a yeast protein. It seems to be part of a peripheral membrane protein complex localized on cis/medial Golgi cisternae where it may participate in tethering intra-Golgi transport vesicles.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CREB-regulated transcription coactivator 3; TORC-3; transducer of regulated cAMP response element-binding protein (CREB) 3; Transducer of regulated cAMP response element-binding protein 3; transducer of regulated CREB protein 3
Gene Aliases: CRTC3; TORC-3; TORC3
UniProt ID: (Human) Q6UUV7
Entrez Gene ID: (Human) 64784
Molecular Function:
transcription cofactor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support