Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
TFCP2L1 is a candidate CP2 family member. It is expressed in a developmentally regulated fashion in vivo and acts as a direct repressor of transcription. CP2-related proteins comprise a family of DNA-binding transcription factors that are generally activators of transcription and expressed ubiquitously.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CP2-related transcriptional repressor 1; CRTR-1; Tcfcp2-related transcriptional repressor 1; Transcription factor CP2-like protein 1; Transcription factor LBP-9; transcription repressor CRTR-1
Gene Aliases: 1810030F05Rik; 4932442M07Rik; AA575098; Cp2l1; Crtr-1; CRTR1; D930018N21Rik; LBP-9; LBP9; Tcfcp2l1; TFCP2L1
UniProt ID: (Human) Q9NZI6, (Mouse) Q3UNW5
Entrez Gene ID: (Human) 29842, (Mouse) 81879, (Rat) 304741
Molecular Function:
DNA-binding transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support