Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MAATVATAAA VAPAPAPGTD SASSVHWFRK GLRLHDNPAL LAAVRGARCV RCVYILDPWF AASSSVGINR WRFLLQSLED LDTSLRKLNS RLFVVRGQPA DVFPRLFKEW GVTRLTFEYD SEPFGKERDA AIMKMAKEAG VEVVTENSHT LYDLDRIIEL NGQKPPLTYK RFQAIISRME LPKKPVGLVT SQQMESCRAE IQENHDETYG VPSLEELGFP TEGLGPAVWQ GGETEALARL DKHLERKAWV ANYERPRMNA NSLLASPTGL SPYLRFGCLS CRLFYYRLWD LYKKVKRNST PPLSLFGQLL WREFFYTAAT NNPRFDRMEG NPICIQIPWD RNPEALAKWA EGKTGFPWID AIMTQLRQEG WIHHLARHAV ACFLTRGDLW VSWESGVRVF DELLLDADFS VNAGSWMWLS CSAFFQQFFH CYCPVGFGRR TDPSGDYIRR YLPKLKAFPS RYIYEPWNAP ESIQKAAKCI IGVDYPRPIV NHAETSRLNI ERMKQIYQQL SRYRGLCLLA SVPSCVEDLS HPVAEPSSSQ AGSMSSAGPR PLPSGPASPK RKLEAAEEPP GEELSKRARV AELPTPELPS KDA
Various biochemical, physiological and behavioral processes display circadian rhythms controlled by an internal biological clock. The central "gears" driving this clock appear to be composed of an autoregulatory transcription/post translation-based feedback loop. Cryptochrome 1 (CRY1) and 2 (CRY2) are DNA-binding flavoproteins that bear some homology to blue-light receptors and photolyases. In Drosophila, CRY is a photoreceptor for the circadian clock where it binds to the clock component TIM in a light-dependent fashion and blocks its function. Mammalian CRY1 and CRY2 function via light-independent interactions with circadian genes CLOCK and BMAL1, as well as with PER1, PER2, and TIM. They seem to act as light-independent components of the circadian clock and likely regulate Per1 transcriptional cycling via interactions with both the activator and its feedback inhibitors. Mutant mice not expressing the Cry1 or Cry2 protein display accelerated and delayed periodicity of locomotor activity, respectively. It appears that the combination of both proteins working together is essential to synchronize the organism to circadian phases. A critical balance between Cry1 and Cry2 is required for proper clock function; in complete darkness, double-mutant mice present with instantaneous arrhythmicity, indicating the absence of an internal circadian clock.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: cryptochrome 2 (photolyase-like); Cryptochrome-2; growth-inhibiting protein 37
Gene Aliases: CRY2; HCRY2; KIAA0658; PHLL2
UniProt ID: (Human) Q49AN0
Entrez Gene ID: (Human) 1408
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support