Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MSKTGTKITF YEDKNFQGRR YDCDCDCADF HTYLSRCNSI KVEGGTWAVY ERPNFAGYMY ILPQGEYPEY QRWMGLNDRL SSCRAVHLPS GGQYKIQIFE KGDFSGQMYE TTEDCPSIME QFHMREIHSC KVLEGVWIFY ELPNYRGRQY LLDKKEYRKP IDWGAASPAV QSFRRIVE
Crystallins are separated into two classes: taxon-specific, or enzyme, and ubiquitous. The latter class constitutes the major proteins of vertebrate eye lens and maintains the transparency and refractive index of the lens. Since lens central fiber cells lose their nuclei during development, these crystallins are made and then retained throughout life, making them extremely stable proteins. Mammalian lens crystallins are divided into alpha, beta, and gamma families; beta and gamma crystallins are also considered as a superfamily. Alpha and beta families are further divided into acidic and basic groups. Seven protein regions exist in crystallins: four homologous motifs, a connecting peptide, and N- and C-terminal extensions. Gamma-crystallins are a homogeneous group of highly symmetrical, monomeric proteins typically lacking connecting peptides and terminal extensions. They are differentially regulated after early development. This gene encodes a protein initially considered to be a beta-crystallin but the encoded protein is monomeric and has greater sequence similarity to other gamma-crystallins. This gene encodes the most significant gamma-crystallin in adult eye lens tissue. Whether due to aging or mutations in specific genes, gamma-crystallins have been involved in cataract formation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Beta-crystallin S; crystallin, gamma 8; crystallin, gamma S; Gamma-crystallin S; Gamma-S-crystallin
Gene Aliases: CRYG8; CRYGS; CTRCT20
UniProt ID: (Human) P22914, (Rat) P0C5E9
Entrez Gene ID: (Human) 1427, (Rat) 689897
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support