Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 4
PA1-776 detects Cysteine String Protein Alpha in mouse and rat brain samples.
PA1-776 has been successfully used in Western blot procedures. By Western blot, this antibody detects a ~34 kDa protein representing Cysteine String Protein Alpha in rat and mouse brain homogenates.
The PA1-776 immunogen is a synthetic peptide corresponding to residues E(180) T T Q L T A D S H P S Y H T D G F N(198) of rat CSP. This sequence is conserved across many species including mouse, human and bovine. This peptide (Cat. # PEP-287) is available for use in neutralization and control experiments.
Cysteine string protein alpha (CSPalpha) is a synaptic vesicle protein that contains a DNA-J domain characteristic of Hsp40 chaperones. Cysteine-string proteins are membrane-bound proteins associated primarily with nerve endings and synaptic vesicles in the brain. These proteins play an important role in presynaptic function and may be involved in calcium-dependent neurotransmitter release. CSP proteins may act as presynaptic chaperones that assist in maintaining synaptic function.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Ceroid-lipofuscinosis neuronal protein 4; CSP; CSP alpha; Cysteine string protein; cysteine string protein alpha; Cysteine-string protein isoform alpha; DnaJ (Hsp40) homolog, subfamily C, member 5; DnaJ homolog subfamily C member 5
Gene Aliases: 2610314I24Rik; AU018536; CLN4; CLN4B; CSP; Cspalpha; DNAJC5; DNAJC5A; mir-941-2; mir-941-3; mir-941-4; mir-941-5; NCL
UniProt ID: (Human) Q9H3Z4, (Rat) P60905, (Mouse) P60904
Entrez Gene ID: (Human) 80331, (Rat) 79130, (Mouse) 13002
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support