Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Peptide Sequence: LWEISTGRTL VRYTGAGLSG RQVHRTQAVF NHTEDYVLLP DERTISLCCW DSRTAERRNL LSLGHNNIVR CIVHSPTNPG FMTCSDDFRA RFWYRRSTTD
This gene encodes one of three subunits which combine to form cleavage stimulation factor (CSTF). CSTF is involved in the polyadenylation and 3'end cleavage of pre-mRNAs. Similar to mammalian G protein beta subunits, this protein contains transducin-like repeats. Several transcript variants with different 5' UTR, but encoding the same protein, have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CF-1 50 kDa subunit; Cleavage stimulation factor 50 kDa subunit; Cleavage stimulation factor subunit 1; cleavage stimulation factor, 3' pre-RNA, subunit 1; cleavage stimulation factor, 3' pre-RNA, subunit 1, 50kD; cleavage stimulation factor, 3' pre-RNA, subunit 1, 50kDa; CSTF 50 kDa subunit; CstF50
Gene Aliases: CstF-50; CSTF1; CstFp50
UniProt ID: (Human) Q05048
Entrez Gene ID: (Human) 1477
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support