Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: MAGLTVRDPA VDRSLRSVFV GNIPYEATEG QLKDIFSEVG PVVSFRLVYD RETGKPKGYG FCEYQDQETA LNAMRNLNGR EFSGRALRVD NAASEKNKEE LKSLGTGAPV IESPYGETIS PEDAPESISK AVASLPPEQM FELMKQMKLC VQNSPQEARN MLLQNPQLAY ALLQAQVVMR IVDPEIALKI LHRQTNIPTL IAGNPQPVHG AGPGSGSNVS MNQQNPQAPQ AQSLGGMHVN GAPPLMQASM QGGVPAPGQM PAAVTGPGPG SLAPGGGMQA QVGMPGSGPV SMERGQVPMQ DPRAAMQRGS LPANVPTPRG LLGDAPNDPR GGTLLSVTGE VEPRGYLGPP HQGPPMHHVP GHESRGPPPH ELRGGPLPEP RPLMAEPRGP MLDQRGPPLD GRGGRDPRGI DARGMEARAM EARGLDARGL EARAMEARAM EARAMEARAM EARAMEVRGM EARGMDTRGP VPGPRGPIPS GMQGPSPINM GAVVPQGSRQ VPVMQGTGMQ GASIQGGSQP GGFSPGQNQV TPQDHEKAAL IMQVLQLTAD QIAMLPPEQR QSILILKEQI QKSTGAP
CSTF2 is one of three (including CSTF1 and CSTF3) cleavage stimulation factors which combine to form CSTF which is involved in the polyadenylation and 3'end cleavage of pre-mRNAs. CSTF2 contains a ribonucleoprotein-type RNA binding domain. CSTF2 is upregulated during activation of B cells which results in the switch of IgM heavy chain mRNA from membrane bound form to the secreted form.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: betaCstF-64 variant 2; CF-1 64 kDa subunit; Cleavage stimulation factor 64 kDa subunit; Cleavage stimulation factor subunit 2; cleavage stimulation factor, 3' pre-RNA, subunit 2; cleavage stimulation factor, 3' pre-RNA, subunit 2, 64kD; cleavage stimulation factor, 3' pre-RNA, subunit 2, 64kDa; CstF 64; CSTF 64 kDa subunit
Gene Aliases: CstF-64; CSTF2
UniProt ID: (Human) P33240
Entrez Gene ID: (Human) 1478
Molecular Function: mRNA polyadenylation factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support