Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: QLWRDYNKYE EGINIHLAKK MIEDRSRDYM NARRVAKEYE TVMKGLDRNA PSVPPQNTPQ EAQQVDMWKK YIQWEKSNPL RTEDQTLITK RVMFAYEQCL LVLGHH
Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 99%.
The protein encoded by this gene is one of three (including CSTF1 and CSTF2) cleavage stimulation factors that combine to form the cleavage stimulation factor complex (CSTF). This complex is involved in the polyadenylation and 3' end cleavage of pre-mRNAs. The encoded protein functions as a homodimer and interacts directly with both CSTF1 and CSTF2 in the CSTF complex. Alternative splicing results in multiple transcript variants encoding different isoforms.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CF-1 77 kDa subunit; Cleavage stimulation factor 77 kDa subunit; Cleavage stimulation factor subunit 3; cleavage stimulation factor subunit 3, isoform 1; cleavage stimulation factor, 3' pre-RNA, subunit 3; cleavage stimulation factor, 3' pre-RNA, subunit 3, 77kD; cleavage stimulation factor, 3' pre-RNA, subunit 3, 77kDa; CSTF 77 kDa subunit; MGC117398; MGC43001; MGC75122
Gene Aliases: 4732468G05Rik; C79532; CSTF-77; CSTF3
UniProt ID: (Human) Q12996, (Mouse) Q99LI7
Entrez Gene ID: (Human) 1479, (Mouse) 228410, (Rat) 362178
Molecular Function: RNA metabolism protein mRNA polyadenylation factor RNA processing factor nucleic acid metabolism protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support