Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Recommended positive controls: PC-3, U87-MG, SK-N-SH.
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
CTAGE5/MIA-2 is a secreted cytokine, and a member of the MIA/OTOR family. Members of this family, which also includes MIA, OTOR, and TANGO, share a Src homology-3 (SH3)-like domain. MIA-2 is predominantly expressed in hepatocytes. Elevated levels of MIA-2 may represent a clinically useful marker for diagnosis of hepatic disease activity and severity.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CTAGE family member 5; CTAGE family member 5 ER export factor; CTAGE family, member 5; Cutaneous T-cell lymphoma-associated antigen 5; Melanoma inhibitory activity protein 2; meningioma expressed antigen 6 (coiled-coil proline-rich); Meningioma-expressed antigen 6/11; MIA protein 2; protein cTAGE-5
Gene Aliases: CTAGE5; MEA11; MEA6; MGEA; MGEA11; MGEA6; MIA2
UniProt ID: (Human) Q96PC5
Entrez Gene ID: (Human) 4253
Molecular Function:
membrane traffic protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support